Learn More
Medchemexpress LLC Amino-PEG6-amido-bis-PEG5-N3 | 1147.31 | C48H94N10O21 | 25 MG
SDP
Catalog No. 5000203252
Encompass_Preferred
A cleavable, PEG-based linker containing an azide functional group and a terminal amino group for bioconjugation. Designed as an 11-unit PEG spacer, it provides hydrophilicity and reactive handles for click chemistry and antibody-drug conjugate synthesis.
  • Cleavable 11-unit PEG linker providing hydrophilic spacer.
  • Contains an azide group for copper-catalyzed and strain-promoted click reactions.
  • Terminal amino group enables further conjugation chemistry.
  • Suitable for antibody-drug conjugate and bioconjugation applications.
  • Available in small lab pack sizes for research use.
Catalog No. 50-002-03252 Supplier Medchemexpress LLC Supplier No. HY13095725MG
A cleavable, PEG-based linker containing an azide functional group and a terminal amino group for bioconjugation. Designed as an 11-unit PEG spacer, it provides hydrophilicity and reactive handles for click chemistry and antibody-drug conjugate synthesis.
  • Cleavable 11-unit PEG linker providing hydrophilic spacer.
  • Contains an azide group for copper-catalyzed and strain-promoted click reactions.
  • Terminal amino group enables further conjugation chemistry.
  • Suitable for antibody-drug conjugate and bioconjugation applications.
  • Available in small lab pack sizes for research use.