Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Amino-PEG6-amido-bis-PEG5-N3 | 1147.31 | C48H94N10O21 | 50 MG
SDP

Catalog No. 5000202824
Encompass_Preferred

A cleavable 11-unit polyethylene glycol (PEG) linker containing an azide functional group for click chemistry and bioconjugation applications. Intended for research use only.

  • Cleavable 11-unit PEG linker suitable for antibody-drug conjugates and bioconjugation
  • Contains an azide (N3) functional group for CuAAC and SPAAC click reactions
  • Available in multiple package sizes, including 50 MG
  • Appearance: oil, colorless to light yellow
  • Molecular weight 1147.31; molecular formula C48H94N10O21
  • Store sealed away from moisture and light; in solvent: -80°C (6 months), -20°C (1 month)

Catalog No. 50-002-02824 Supplier Medchemexpress LLC Supplier No. HY13095750MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A cleavable 11-unit polyethylene glycol (PEG) linker containing an azide functional group for click chemistry and bioconjugation applications. Intended for research use only.

  • Cleavable 11-unit PEG linker suitable for antibody-drug conjugates and bioconjugation
  • Contains an azide (N3) functional group for CuAAC and SPAAC click reactions
  • Available in multiple package sizes, including 50 MG
  • Appearance: oil, colorless to light yellow
  • Molecular weight 1147.31; molecular formula C48H94N10O21
  • Store sealed away from moisture and light; in solvent: -80°C (6 months), -20°C (1 month)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.