Learn More
Medchemexpress LLC Amino-PEG6-amine | 76927-70-3 | MFCD30343546 | 97.0% | 324.41 | 250 MG
SDP
Catalog No. 5000402250
Encompass_Preferred
Amino-PEG6-amine is a PROTAC linker, based on a PEG (polyethylene glycol) chain with 6 units, which can be utilized in the synthesis of PROTACs. PROTACs are molecules that contain two different ligands connected by a linker: one ligand binds to an E3 ubiquitin ligase, and the other binds to the target protein. This mechanism allows PROTACs to leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based (6 units) PROTAC linker
  • Can be used in the synthesis of PROTACs
Catalog No. 50-004-02250 Supplier Medchemexpress LLC Supplier No. 250MGHY23105
Amino-PEG6-amine is a PROTAC linker, based on a PEG (polyethylene glycol) chain with 6 units, which can be utilized in the synthesis of PROTACs. PROTACs are molecules that contain two different ligands connected by a linker: one ligand binds to an E3 ubiquitin ligase, and the other binds to the target protein. This mechanism allows PROTACs to leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based (6 units) PROTAC linker
  • Can be used in the synthesis of PROTACs