Learn More
Medchemexpress LLC Amino-PEG7-amine | 332941-25-0 | 99.3% | C16H36N2O7 | 250 MG
SDP
Catalog No. 5000352872
Encompass_Preferred
Amino-PEG7-amine is a PEG-based PROTAC linker used in the synthesis of Proteolysis Targeting Chimeras (PROTACs). These compounds are designed with two ligands connected by a linker: one targeting an E3 ubiquitin ligase and the other for a target protein. PROTACs utilize the intracellular ubiquitin-proteasome system to achieve selective degradation of target proteins. This product is for research use only.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Utilizes the intracellular ubiquitin-proteasome system
  • Designed for selective degradation of target proteins
Catalog No. 50-003-52872 Supplier Medchemexpress LLC Supplier No. 250MGHY140209
Amino-PEG7-amine is a PEG-based PROTAC linker used in the synthesis of Proteolysis Targeting Chimeras (PROTACs). These compounds are designed with two ligands connected by a linker: one targeting an E3 ubiquitin ligase and the other for a target protein. PROTACs utilize the intracellular ubiquitin-proteasome system to achieve selective degradation of target proteins. This product is for research use only.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Utilizes the intracellular ubiquitin-proteasome system
  • Designed for selective degradation of target proteins