Learn More
Medchemexpress LLC Amino-PEG7-amine | 332941-25-0 | MFCD09752883 | 99.3% | C16H36N2O7 | 1 G
SDP
Catalog No. 5000353094
Encompass_Preferred
Amino-PEG7-amine is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs are designed to selectively degrade target proteins by leveraging the intracellular ubiquitin-proteasome system, utilizing two ligands connected by a linker: one targeting an E3 ubiquitin ligase and the other a specific protein.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Facilitates selective protein degradation
  • Applicable in cancer targeted therapy
Catalog No. 50-003-53094 Supplier Medchemexpress LLC Supplier No. HY1402091G
Amino-PEG7-amine is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs are designed to selectively degrade target proteins by leveraging the intracellular ubiquitin-proteasome system, utilizing two ligands connected by a linker: one targeting an E3 ubiquitin ligase and the other a specific protein.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Facilitates selective protein degradation
  • Applicable in cancer targeted therapy