Learn More
Medchemexpress LLC Amino-PEG7-C2-SH hydrochloride | 98.0% | 421.98 | 50 MG
SDP
Catalog No. 5000348635
Encompass_Preferred
Amino-PEG7-C2-SH hydrochloride is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. This mechanism exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Features two ligands connected by a linker
  • One ligand targets an E3 ubiquitin ligase, the other targets a protein
  • Exploits the ubiquitin-proteasome system
  • Enables selective degradation of target proteins
Catalog No. 50-003-48635 Supplier Medchemexpress LLC Supplier No. HY13845150MG
Amino-PEG7-C2-SH hydrochloride is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. This mechanism exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Features two ligands connected by a linker
  • One ligand targets an E3 ubiquitin ligase, the other targets a protein
  • Exploits the ubiquitin-proteasome system
  • Enables selective degradation of target proteins