Learn More
Medchemexpress LLC Amino-PEG8-amine | 82209-36-7 | ≥98.0% | 1 G
SDP
Catalog No. 5000341076
Encompass_Preferred
Amino-PEG8-amine is a PEG-based (8 units) PROTAC linker that can be used in the synthesis of PROTACs. PROTACs consist of two different ligands connected by a linker; one ligand is for an E3 ubiquitin ligase, and the other is for the target protein. PROTACs utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Utilizes the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-41076 Supplier Medchemexpress LLC Supplier No. HY1306591G
Amino-PEG8-amine is a PEG-based (8 units) PROTAC linker that can be used in the synthesis of PROTACs. PROTACs consist of two different ligands connected by a linker; one ligand is for an E3 ubiquitin ligase, and the other is for the target protein. PROTACs utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Utilizes the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins