Learn More
Medchemexpress LLC Amino-PEG8-amine | 82209-36-7 | ≥98.0% | 100 MG
SDP
Catalog No. 5000340233
Encompass_Preferred
Amino-PEG8-amine is a PEG-based (8 units) PROTAC linker used in the synthesis of PROTACs. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PROTAC linker
  • Used in the synthesis of PROTACs
  • Contains two different ligands connected by a linker; one for an E3 ubiquitin ligase and the other for the target protein
  • Exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins
Catalog No. 50-003-40233 Supplier Medchemexpress LLC Supplier No. HY130659100MG
Amino-PEG8-amine is a PEG-based (8 units) PROTAC linker used in the synthesis of PROTACs. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PROTAC linker
  • Used in the synthesis of PROTACs
  • Contains two different ligands connected by a linker; one for an E3 ubiquitin ligase and the other for the target protein
  • Exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins