Learn More
Medchemexpress LLC Amino-PEG8-amine | 82209-36-7 | ≥98.0% | 250 MG
SDP
Catalog No. 5000341969
Encompass_Preferred
Amino-PEG8-amine is a PEG-based (8 units) PROTAC linker used in the synthesis of PROTACs. PROTACs, or Proteolysis-targeting chimeras, consist of two different ligands connected by a linker. One ligand targets an E3 ubiquitin ligase, and the other targets the protein of interest. PROTACs utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based (8 units) linker
  • Can be used in the synthesis of PROTACs
  • Ligands connected by a linker target E3 ubiquitin ligase and protein of interest
  • Utilizes intracellular ubiquitin-proteasome system to selectively degrade target proteins
Catalog No. 50-003-41969 Supplier Medchemexpress LLC Supplier No. 250MGHY130659
Amino-PEG8-amine is a PEG-based (8 units) PROTAC linker used in the synthesis of PROTACs. PROTACs, or Proteolysis-targeting chimeras, consist of two different ligands connected by a linker. One ligand targets an E3 ubiquitin ligase, and the other targets the protein of interest. PROTACs utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based (8 units) linker
  • Can be used in the synthesis of PROTACs
  • Ligands connected by a linker target E3 ubiquitin ligase and protein of interest
  • Utilizes intracellular ubiquitin-proteasome system to selectively degrade target proteins