Learn More
Medchemexpress LLC Amino-PEG8-amine | 82209-36-7 | 98.0% | 5 G
SDP
Catalog No. 5000338196
Encompass_Preferred
Amino-PEG8-amine is a PEG-based (8 units) PROTAC linker that is used in the synthesis of PROTACs. PROTACs are compounds that contain two different ligands connected by a linker; one ligand is for an E3 ubiquitin ligase and the other is for the target protein. These PROTACs leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PROTACs contain two different ligands connected by a linker.
  • One ligand is for an E3 ubiquitin ligase.
  • The other ligand is for the target protein.
  • They exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
Catalog No. 50-003-38196 Supplier Medchemexpress LLC Supplier No. HY1306595G
Amino-PEG8-amine is a PEG-based (8 units) PROTAC linker that is used in the synthesis of PROTACs. PROTACs are compounds that contain two different ligands connected by a linker; one ligand is for an E3 ubiquitin ligase and the other is for the target protein. These PROTACs leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PROTACs contain two different ligands connected by a linker.
  • One ligand is for an E3 ubiquitin ligase.
  • The other ligand is for the target protein.
  • They exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.