Learn More
Medchemexpress LLC Amino-PEG8-Boc | 756526-06-4 | 99.8% | 497.62 | C23H47NO10 | 250 MG
SDP
Catalog No. 5000203597
Encompass_Preferred
Amino-PEG8-Boc is an eight-unit polyethylene glycol (PEG) linker bearing a Boc-protected terminal amine. It is used as a cleavable linker in the synthesis of antibody-drug conjugates (ADCs) and as a PEG-based linker for PROTACs and other bioconjugation chemistries.
  • Cleavable 8-unit PEG linker
  • Protecting group: Boc on terminal amine
  • Suitable for antibody-drug conjugate synthesis
  • Suitable for PROTAC linker construction
  • Available as 250 mg research pack
  • High purity for research use
Catalog No. 50-002-03597 Supplier Medchemexpress LLC Supplier No. HYW019799250MG
Amino-PEG8-Boc is an eight-unit polyethylene glycol (PEG) linker bearing a Boc-protected terminal amine. It is used as a cleavable linker in the synthesis of antibody-drug conjugates (ADCs) and as a PEG-based linker for PROTACs and other bioconjugation chemistries.
  • Cleavable 8-unit PEG linker
  • Protecting group: Boc on terminal amine
  • Suitable for antibody-drug conjugate synthesis
  • Suitable for PROTAC linker construction
  • Available as 250 mg research pack
  • High purity for research use