Learn More
Medchemexpress LLC Amino-PEG9-acid | 1191079-83-0 | 485.57 | C21H43NO11 | 1 G
SDP
Catalog No. 5000203806
Encompass_Preferred
Amino-PEG9-acid is a polyethylene glycol (PEG)-based linker with a terminal amino group and a carboxylic acid, used as a PROTAC linker and as a non-cleavable ADC linker for antibody-drug conjugates. It provides a hydrophilic spacer useful for conjugation chemistry in bioconjugation and linker design.
  • Is a PEG-based PROTAC linker.
  • Can be used in the synthesis of PROTACs.
  • Functions as a non-cleavable ADC linker for antibody-drug conjugates.
  • Provides a hydrophilic spacer with terminal amino and carboxylic acid groups.
  • Available in multiple package sizes, including 1 g for lab-scale work.
Catalog No. 50-002-03806 Supplier Medchemexpress LLC Supplier No. HY1301661G
Amino-PEG9-acid is a polyethylene glycol (PEG)-based linker with a terminal amino group and a carboxylic acid, used as a PROTAC linker and as a non-cleavable ADC linker for antibody-drug conjugates. It provides a hydrophilic spacer useful for conjugation chemistry in bioconjugation and linker design.
  • Is a PEG-based PROTAC linker.
  • Can be used in the synthesis of PROTACs.
  • Functions as a non-cleavable ADC linker for antibody-drug conjugates.
  • Provides a hydrophilic spacer with terminal amino and carboxylic acid groups.
  • Available in multiple package sizes, including 1 g for lab-scale work.