Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Amino-PEG9-amine | 474082-35-4 | >98% | C20H44N2O9 | 250 MG
SDP

Catalog No. 5000352873
Encompass_Preferred

Amino-PEG9-amine is a PEG-based PROTAC linker used in the synthesis of PROTACs. These compounds employ the intracellular ubiquitin-proteasome system to selectively degrade target proteins. This mechanism involves two distinct ligands, connected by a linker, where one ligand targets an E3 ubiquitin ligase and the other binds to the protein of interest.

  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Applicable in targeted therapy drug development
  • For research use only
  • Molecular weight of 456.57
  • Store at 4°C, protect from light. In solvent: store at -80°C for 6 months, or -20°C for 1 month (protect from light).

Catalog No. 50-003-52873 Supplier Medchemexpress LLC Supplier No. 250MGHY140210
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Amino-PEG9-amine is a PEG-based PROTAC linker used in the synthesis of PROTACs. These compounds employ the intracellular ubiquitin-proteasome system to selectively degrade target proteins. This mechanism involves two distinct ligands, connected by a linker, where one ligand targets an E3 ubiquitin ligase and the other binds to the protein of interest.

  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Applicable in targeted therapy drug development
  • For research use only
  • Molecular weight of 456.57
  • Store at 4°C, protect from light. In solvent: store at -80°C for 6 months, or -20°C for 1 month (protect from light).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.