Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Amino-PEG9-amine | 474082-35-4 | C20H44N2O9 | 500 MG
SDP

Catalog No. 5000357324
Encompass_Preferred

Amino-PEG9-amine is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs are compounds that leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins, featuring two distinct ligands connected by a linker: one for an E3 ubiquitin ligase and the other for the target protein.

  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • Molecular weight: 456.57
  • Appearance: Liquid
  • Color: Colorless to light yellow

Catalog No. 50-003-57324 Supplier Medchemexpress LLC Supplier No. HY140210500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Amino-PEG9-amine is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs are compounds that leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins, featuring two distinct ligands connected by a linker: one for an E3 ubiquitin ligase and the other for the target protein.

  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • Molecular weight: 456.57
  • Appearance: Liquid
  • Color: Colorless to light yellow

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.