Learn More
Medchemexpress LLC Amino-PEG9-Boc | 1818294-44-8 | C25H51NO11 | 100 MG
SDP
Catalog No. 5000357214
Encompass_Preferred
Amino-PEG9-Boc is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Functions as a PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Leverages the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-57214 Supplier Medchemexpress LLC Supplier No. HY140195100MG
Amino-PEG9-Boc is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Functions as a PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Leverages the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins