Learn More
Medchemexpress LLC Amino Tadalafil | 25MG
SDP
Catalog No. 5000229219
Encompass_Preferred
Amino Tadalafil is a structural analogue of Tadalafil (HY-90009A) Tadalafil is a potent inhibitor of PDE5 with applications in several conditions including erectile dysfunction pulmonary arterial hypertension and lower urinary tract dysfunction[1][2]
Catalog No. 50-002-29219 Supplier Medchemexpress LLC Supplier No. HY11710925MG
Amino Tadalafil is a structural analogue of Tadalafil (HY-90009A) Tadalafil is a potent inhibitor of PDE5 with applications in several conditions including erectile dysfunction pulmonary arterial hypertension and lower urinary tract dysfunction[1][2]