Learn More
Apexbio Technology LLC Amino Tadalafil 385769-84-6 100mg
SDP
Supplier:  Apexbio Technology LLC C3266100
Encompass_Preferred
Amino Tadalafil (CAS 385769-84-6) is a small molecule that functions as a potent inhibitor of phosphodiesterase 5 (PDE5) an enzyme primarily expressed in the smooth muscle of the corpus cavernosum By structurally mimicking cGMP Amino Tadalafil competitively binds to PDE5 preventing the hydrolysis of cGMP and thereby enhancing nitric oxide-mediated vasodilation Preclinical studies have shown that Amino Tadalafil a structural analogue of tadalafil may be relevant for investigating mechanisms of erectile function and dysfunction It is also utilized in research evaluating PDE5 inhibitor adulteration in supplements and in animal models addressing erectile dysfunction associated with oxidative stress and spinal cord injury
Catalog No. 50-000-89144