Learn More
Sigma Aldrich Fine Chemicals Biosciences Amino terminal fLAG BAPTM
Catalog No. 502904375
Encompass
The Amino-terminal fLAG-BAP fusion Protein is a control prorein used to confirm the funtional integrity of the anti-fLAG M1 and M2 monoclonal antibodies in applications such as western blotting immunoprecipitation ELISA electron microscopy and fACS.
Catalog No. 50-290-4375 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. P7582100UG
The Amino-terminal fLAG-BAP fusion Protein is a control prorein used to confirm the funtional integrity of the anti-fLAG M1 and M2 monoclonal antibodies in applications such as western blotting immunoprecipitation ELISA electron microscopy and fACS.