Learn More
Medchemexpress LLC Amino-tri-(carboxyethoxymethyl)-methane hydrochloride | 1416771-72-6 | MFCD34622747 | 97.0% | 373.78 g/mol | C13H24ClNO9 | 25 MG
SDP
Catalog No. 5000227644
Encompass_Preferred
Amino-Tri-(carboxyethoxymethyl)-methane hydrochloride is a PEG-based cleavable linker used in the synthesis of antibody-drug conjugates (ADCs) and PROTACs. Supplied as the hydrochloride salt, the reagent is provided as a white to off-white solid in small research-scale quantities and intended for research use only.
  • Cleavable PEG linker suitable for ADC and PROTAC synthesis.
  • High purity (97.0%) for reliable research results.
  • Molecular weight 373.78 g/mol and formula C13H24ClNO9.
  • Available in small pack sizes for lab-scale workflows, such as 25 MG.
  • Solid, white to off-white appearance; store sealed away from moisture and light.
  • For research use only; not for clinical or human use.
Catalog No. 50-002-27644 Supplier Medchemexpress LLC Supplier No. HY117519A25MG
Amino-Tri-(carboxyethoxymethyl)-methane hydrochloride is a PEG-based cleavable linker used in the synthesis of antibody-drug conjugates (ADCs) and PROTACs. Supplied as the hydrochloride salt, the reagent is provided as a white to off-white solid in small research-scale quantities and intended for research use only.
  • Cleavable PEG linker suitable for ADC and PROTAC synthesis.
  • High purity (97.0%) for reliable research results.
  • Molecular weight 373.78 g/mol and formula C13H24ClNO9.
  • Available in small pack sizes for lab-scale workflows, such as 25 MG.
  • Solid, white to off-white appearance; store sealed away from moisture and light.
  • For research use only; not for clinical or human use.