Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Aminoacetone hydrochloride | 7737-17-9 | 109.55 | 10 G
SDP

Catalog No. 5000449682
Encompass_Preferred

Aminoacetone hydrochloride is the simplest monopeptide, serving as an intermediate in the metabolism of threonine and glycine. This compound is also an endogenous substrate for semicarbazide-sensitive amine oxidase (SSAO) and can be used for the determination of SSAO activity.

  • Endogenous substrate for semicarbazide-sensitive amine oxidase (SSAO)
  • Can be utilized for determining SSAO activity
  • Appearance: Light yellow to yellow solid
  • Molecular formula: C3H8ClNO
  • Soluble in DMSO: 200 mg/mL

Catalog No. 50-004-49682 Supplier Medchemexpress LLC Supplier No. HYW00485010G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Aminoacetone hydrochloride is the simplest monopeptide, serving as an intermediate in the metabolism of threonine and glycine. This compound is also an endogenous substrate for semicarbazide-sensitive amine oxidase (SSAO) and can be used for the determination of SSAO activity.

  • Endogenous substrate for semicarbazide-sensitive amine oxidase (SSAO)
  • Can be utilized for determining SSAO activity
  • Appearance: Light yellow to yellow solid
  • Molecular formula: C3H8ClNO
  • Soluble in DMSO: 200 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.