Learn More
Medchemexpress LLC Aminoacetone hydrochloride | 7737-17-9 | 109.55 | 25 G
SDP
Catalog No. 5000448006
Encompass_Preferred
Aminoacetone hydrochloride is identified as the simplest monopeptide and acts as an intermediate in the metabolic pathways of threonine and glycine. It serves as an endogenous substrate for semicarbazide-sensitive amine oxidase (SSAO) and can be used for determining SSAO activity.
  • Simplest monopeptide
  • Intermediate in threonine and glycine metabolism
  • Endogenous substrate for semicarbazide-sensitive amine oxidase (SSAO)
  • Can be used for determination of SSAO activity
Catalog No. 50-004-48006 Supplier Medchemexpress LLC Supplier No. 25GHYW004850
Aminoacetone hydrochloride is identified as the simplest monopeptide and acts as an intermediate in the metabolic pathways of threonine and glycine. It serves as an endogenous substrate for semicarbazide-sensitive amine oxidase (SSAO) and can be used for determining SSAO activity.
  • Simplest monopeptide
  • Intermediate in threonine and glycine metabolism
  • Endogenous substrate for semicarbazide-sensitive amine oxidase (SSAO)
  • Can be used for determination of SSAO activity