Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aminoacetonitrile HCl, MP Biomedicals

Catalog No. ICN225879 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN225879 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN225879 Supplier MP Biomedicals, Inc Supplier No. 0522587950

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6011-14-9
Molecular Formula C2H5ClN2
Molecular Weight (g/mol) 92.526
InChI Key XFKYKTBPRBZDFG-UHFFFAOYSA-N
Synonym aminoacetonitrile hydrochloride, 2-aminoacetonitrile hydrochloride, glycinonitrile hydrochloride, aminoacetonitrile hcl, unii-s8l3v1ksb7, acetonitrile, amino-, monohydrochloride, s8l3v1ksb7, aminoacetonitrile.hcl, amino-acetonitrile hcl, acmc-209mhz
PubChem CID 11491918
IUPAC Name 2-aminoacetonitrile;hydrochloride
SMILES C(C#N)N.Cl

Specifications

Quantity 50 mg
Formula Weight 92.53
Chemical Name or Material Amino Acetonitrile HCl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.