Learn More
PREDICT HEALTH Disorder: Metabolic disorders related with Amino acid pathway Parameters: : 47 Amino AcidsAnalysis Method: LC-MS/MS
Supplier:  PREDICT HEALTH BR130030
Encompass_Preferred
Intended Use: Confirmation & Monitoring of metabolic disorders Disorder: Metabolic disorders related with Amino acid pathway Parameters: : 47 Amino AcidsAnalysis Method: LC-MS/MS Evaluation: QuantitativeSample Type: Plasma, Urine and CSF
Catalog No. NC2891047