Learn More
Medchemexpress LLC HY-108939 1mg Medchemexpress, Aminoacyl tRNA synthetase-IN-1 CAS:219931-45-0 Purity:>98%
SDP
Catalog No. 501968635
Encompass_Preferred
Medchemexpress, HY-108939 1mg Aminoacyl tRNA synthetase-IN-1 CAS:219931-45-0 Aminoacyl tRNA synthetase-IN-1 is a bacterial aminoacyl tRNA synthetase (aaRS) inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8635 Supplier Medchemexpress LLC Supplier No. HY1089391MG
Medchemexpress, HY-108939 1mg Aminoacyl tRNA synthetase-IN-1 CAS:219931-45-0 Aminoacyl tRNA synthetase-IN-1 is a bacterial aminoacyl tRNA synthetase (aaRS) inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.