Learn More
Medchemexpress LLC Aminoacyl tRNA synthetase-IN-1 | 219931-45-0 | MFCD31619268 | 99.8% | 459.48 g/mol | C16H25N7O7S | 5 MG
SDP
Catalog No. 5000263593
Encompass_Preferred
Aminoacyl tRNA synthetase-IN-1 is a research-grade bacterial aminoacyl-tRNA synthetase (aaRS) inhibitor reported to inhibit Staphylococcus aureus, Enterococcus faecalis, and Escherichia coli. Key chemical identifiers include CAS 219931-45-0, molecular formula C16H25N7O7S, and molecular weight 459.48 g/mol. The material is supplied as a small-mass solid intended for in vitro laboratory research.
  • Inhibits bacterial aminoacyl-tRNA synthetase enzymes.
  • Reported activity against S. aureus, E. faecalis, and E. coli.
  • High purity suitable for research use (99.8%).
  • Supplied in small milligram quantities suitable for biochemical assays.
  • Useful for studying bacterial translation and antimicrobial mechanisms.
  • Provided with supporting analytical data such as COA and HPLC.
Catalog No. 50-002-63593 Supplier Medchemexpress LLC Supplier No. HY1089395MG
Aminoacyl tRNA synthetase-IN-1 is a research-grade bacterial aminoacyl-tRNA synthetase (aaRS) inhibitor reported to inhibit Staphylococcus aureus, Enterococcus faecalis, and Escherichia coli. Key chemical identifiers include CAS 219931-45-0, molecular formula C16H25N7O7S, and molecular weight 459.48 g/mol. The material is supplied as a small-mass solid intended for in vitro laboratory research.
  • Inhibits bacterial aminoacyl-tRNA synthetase enzymes.
  • Reported activity against S. aureus, E. faecalis, and E. coli.
  • High purity suitable for research use (99.8%).
  • Supplied in small milligram quantities suitable for biochemical assays.
  • Useful for studying bacterial translation and antimicrobial mechanisms.
  • Provided with supporting analytical data such as COA and HPLC.