Learn More
Medchemexpress LLC HY-113328 1g Medchemexpress, Aminoadipic acid CAS:542-32-5 Purity:>98%
SDP
Catalog No. 501932460
Encompass_Preferred
Medchemexpress, HY-113328 1g Aminoadipic acid CAS:542-32-5 Aminoadipic acid is an intermediate in the metabolism of lysine and saccharopine. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2460 Supplier Medchemexpress LLC Supplier No. HY1133281G
Medchemexpress, HY-113328 1g Aminoadipic acid CAS:542-32-5 Aminoadipic acid is an intermediate in the metabolism of lysine and saccharopine. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.