Learn More
Medchemexpress LLC Aminoadipic acid-d3 | 2483832-01-3 | 99.1% | 164.17 | 10 MG
SDP
Catalog No. 5000437068
Encompass_Preferred
Aminoadipic acid-d3 is the deuterium labeled Aminoadipic acid, an intermediate in the metabolism of lysine and saccharopine. Deuterium substitution has the potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-37068 Supplier Medchemexpress LLC Supplier No. HY113328S10MG
Aminoadipic acid-d3 is the deuterium labeled Aminoadipic acid, an intermediate in the metabolism of lysine and saccharopine. Deuterium substitution has the potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.