Learn More
Medchemexpress LLC Aminoadipic acid-d3 | 2483832-01-3 | MFCD06658660 | 99.1% | 164.17 | C6H8D3NO4 | 5 MG
SDP
Catalog No. 5000264241
Encompass_Preferred
Aminoadipic acid-d3 is the deuterium-labeled isotopologue of aminoadipic acid (CAS 2483832-01-3), supplied as a high-purity reference standard for analytical and metabolic research. The d3 label permits precise tracking and quantitation in stable isotope tracing and LC-MS workflows, and the compound is provided in small, accurately weighed packages for use as an internal standard.
Catalog No. 50-002-64241 Supplier Medchemexpress LLC Supplier No. HY113328S5MG
Aminoadipic acid-d3 is the deuterium-labeled isotopologue of aminoadipic acid (CAS 2483832-01-3), supplied as a high-purity reference standard for analytical and metabolic research. The d3 label permits precise tracking and quantitation in stable isotope tracing and LC-MS workflows, and the compound is provided in small, accurately weighed packages for use as an internal standard.