Learn More
Apexbio Technology LLC Aminoallyl-dUTP(Synonyms: 5-(3-Aminoallyl)-2'-deoxyuridine-5'-triphosphate, AA-dUTP, Aminoallyl-2'-deoxyuridine triphosphate), 100ul (10mM), CAS: 116840-18-7.
SDP
Catalog No. 5000061238
Encompass_Preferred
Aminoallyl-dUTP (CAS 116840-18-7) is a modified deoxynucleotide designed for indirect covalent labeling of DNA or cDNA in molecular biology research Structurally analogous to natural dTTP it incorporates into DNA via enzymatic reactions including PCR (Taq polymerase) nick translation primer extension (Klenow exo-) terminal labeling (TdT) and reverse transcription (MMLV RT) The introduced primary amine moiety enables subsequent conjugation to NHS-activated labels such as fluorophores for fluorescence microscopy or biotin for affinity-based purification and detection strategies including in situ hybridization
Catalog No. 50-000-61238 Supplier Apexbio Technology LLC Supplier No. B79500.1
Aminoallyl-dUTP (CAS 116840-18-7) is a modified deoxynucleotide designed for indirect covalent labeling of DNA or cDNA in molecular biology research Structurally analogous to natural dTTP it incorporates into DNA via enzymatic reactions including PCR (Taq polymerase) nick translation primer extension (Klenow exo-) terminal labeling (TdT) and reverse transcription (MMLV RT) The introduced primary amine moiety enables subsequent conjugation to NHS-activated labels such as fluorophores for fluorescence microscopy or biotin for affinity-based purification and detection strategies including in situ hybridization