Learn More
Apexbio Technology LLC Aminoallyl-dUTP-X-Cy5(Synonyms: AA-dUTP-Cy5, Aminoallyl-deoxyuridine triphosphate-Cy5, Cy5-AA-dUTP, Cy5-labeled aminoallyl-dUTP), 10ul (5mM).
SDP
Catalog No. 5000060356
Encompass_Preferred
Aminoallyl-dUTP-X-Cy5 is a fluorescently labeled deoxynucleotide analogue structurally resembling natural thymidine triphosphate (dTTP) It serves as a substrate suitable for enzymatic incorporation into DNA strands through methods such as PCR reverse transcription nick translation or random priming Upon incorporation this molecule confers red fluorescence (excitation/emission 650/665 nm) to nucleic acid probes facilitating their detection in applications including in situ hybridization microarray assays blotting procedures and multiplex fluorescence analyses
Catalog No. 50-000-60356 Supplier Apexbio Technology LLC Supplier No. B88190.01
Aminoallyl-dUTP-X-Cy5 is a fluorescently labeled deoxynucleotide analogue structurally resembling natural thymidine triphosphate (dTTP) It serves as a substrate suitable for enzymatic incorporation into DNA strands through methods such as PCR reverse transcription nick translation or random priming Upon incorporation this molecule confers red fluorescence (excitation/emission 650/665 nm) to nucleic acid probes facilitating their detection in applications including in situ hybridization microarray assays blotting procedures and multiplex fluorescence analyses