Learn More
Apexbio Technology LLC Aminoallyl-dUTP-X-Cy5(Synonyms: AA-dUTP-Cy5, Aminoallyl-deoxyuridine triphosphate-Cy5, Cy5-AA-dUTP, Cy5-labeled aminoallyl-dUTP), 5X10ul (5mM).
SDP
Catalog No. 5000060402
Encompass_Preferred
Aminoallyl-dUTP-X-Cy5 is a fluorescently labeled deoxynucleotide analogue structurally resembling natural thymidine triphosphate (dTTP). It serves as a substrate suitable for enzymatic incorporation into DNA strands through methods such as PCR, reverse transcription, nick translation, or random priming. Upon incorporation, this molecule confers red fluorescence (excitation/emission: 650/665 nm) to nucleic acid probes, facilitating their detection in applications including in situ hybridization, microarray assays, blotting procedures, and multiplex fluorescence analyses.
Catalog No. 50-000-60402 Supplier Apexbio Technology LLC Supplier No. B88190.05
Aminoallyl-dUTP-X-Cy5 is a fluorescently labeled deoxynucleotide analogue structurally resembling natural thymidine triphosphate (dTTP). It serves as a substrate suitable for enzymatic incorporation into DNA strands through methods such as PCR, reverse transcription, nick translation, or random priming. Upon incorporation, this molecule confers red fluorescence (excitation/emission: 650/665 nm) to nucleic acid probes, facilitating their detection in applications including in situ hybridization, microarray assays, blotting procedures, and multiplex fluorescence analyses.