Learn More
Apexbio Technology LLC Aminoallyl-UTP 1ml (10mM)
SDP
Catalog No. 503107120
Encompass_Preferred
Aminoallyl-UTP is used for Nucleic Acid Sequence-Based Amplification (NASBA) gene chip-based hybridization detection Eberwine mRNA amplification and RNA labeling During NASBA reactions and Eberwine mRNA amplification processes Aminoallyl-UTP is incorporated into RNA sequences The resulting amino-functionalized RNA molecules can then be conjugated with fluorescent groups via primary amine/NHS ester chemistry (using NHS ester of fluorescent dyes) for subsequent gene chip-based hybridization detection
Catalog No. 50-310-7120 Supplier Apexbio Technology LLC Supplier No. B79510.1
Aminoallyl-UTP is used for Nucleic Acid Sequence-Based Amplification (NASBA) gene chip-based hybridization detection Eberwine mRNA amplification and RNA labeling During NASBA reactions and Eberwine mRNA amplification processes Aminoallyl-UTP is incorporated into RNA sequences The resulting amino-functionalized RNA molecules can then be conjugated with fluorescent groups via primary amine/NHS ester chemistry (using NHS ester of fluorescent dyes) for subsequent gene chip-based hybridization detection