Learn More
Apexbio Technology LLC Aminoallyl-UTP 100ul (10mM)
SDP
Catalog No. 502240228
Encompass_Preferred
Aminoallyl-UTP is used for Nucleic Acid Sequence-Based Amplification (NASBA) gene chip-based hybridization detection Eberwine mRNA amplification and RNA labeling During NASBA reactions and Eberwine mRNA amplification processes Aminoallyl-UTP is incorporated into RNA sequences The resulting amino-functionalized RNA molecules can then be conjugated with fluorescent groups via primary amine/NHS ester chemistry (using NHS ester of fluorescent dyes) for subsequent gene chip-based hybridization detection
Catalog No. 50-224-0228 Supplier Apexbio Technology LLC Supplier No. B79515.1
Aminoallyl-UTP is used for Nucleic Acid Sequence-Based Amplification (NASBA) gene chip-based hybridization detection Eberwine mRNA amplification and RNA labeling During NASBA reactions and Eberwine mRNA amplification processes Aminoallyl-UTP is incorporated into RNA sequences The resulting amino-functionalized RNA molecules can then be conjugated with fluorescent groups via primary amine/NHS ester chemistry (using NHS ester of fluorescent dyes) for subsequent gene chip-based hybridization detection