Learn More
Apexbio Technology LLC Aminoallyl-UTP-X - Cy3 10ul (5mM)
SDP
Catalog No. 5000088673
Encompass_Preferred
Aminoallyl-UTP-X - Cy3 is a modified nucleotide analog designed for fluorescent labeling of RNA molecules Structurally it is composed of uridine triphosphate (UTP) conjugated to a Cy3 fluorophore via an aminoallyl linker enabling its incorporation during in vitro transcription reactions Once integrated into RNA transcripts Cy3-labeled RNA can be visualized by standard fluorescence imaging techniques Based on these properties Aminoallyl-UTP-X - Cy3 is applicable in RNA visualization RNA localization studies RNA-protein interaction assays and fluorescence-based RNA detection in biomedical research
Catalog No. 50-000-88673 Supplier Apexbio Technology LLC Supplier No. B83310.01
Aminoallyl-UTP-X - Cy3 is a modified nucleotide analog designed for fluorescent labeling of RNA molecules Structurally it is composed of uridine triphosphate (UTP) conjugated to a Cy3 fluorophore via an aminoallyl linker enabling its incorporation during in vitro transcription reactions Once integrated into RNA transcripts Cy3-labeled RNA can be visualized by standard fluorescence imaging techniques Based on these properties Aminoallyl-UTP-X - Cy3 is applicable in RNA visualization RNA localization studies RNA-protein interaction assays and fluorescence-based RNA detection in biomedical research