Learn More
Medchemexpress LLC Aminobenzenesulfonic auristatin E-d8 | 00-00-0 | MFCD34676922 | 98.0% | 761.05 | C37H56D8N6O8S | 5 MG

Supplier: Medchemexpress LLC HY145989S5MG
Aminobenzenesulfonic auristatin E-d8 is a deuterated drug-linker conjugate supplied as a white to off-white solid for research use. It serves as an auristatin E payload bearing an aminobenzenesulfonic moiety and is intended for antibody-drug conjugate (ADC) synthesis and related preclinical studies.
- Purity 98.0%
- Molecular weight 761.05
- Chemical formula C37H56D8N6O8S
- White to off-white solid appearance
- Suitable for ADC synthesis and preclinical research
- Isotopically labeled (d8) for mass spectrometry tracing
- Provided as a small-scale research quantity (5 MG)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.