Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Aminobenzenesulfonic auristatin E-d8 | 00-00-0 | MFCD34676922 | 98.0% | C37H56D8N6O8S | 10MG
SDP

Catalog No. 5000258373
Encompass_Preferred

Aminobenzenesulfonic auristatin E-d8 is a deuterium-labeled drug-linker reagent used in antibody-drug conjugate (ADC) research. It combines auristatin E with an aminobenzenesulfonic linker and is supplied as a solid for preclinical, analytical, and linker-optimization studies. It is intended for research use only.

  • Deuterium-labeled version of auristatin E for tracer and stability studies.
  • High purity (98.0%).
  • Chemical formula: C37H56D8N6O8S.
  • Molecular weight: 761.05.
  • White to off-white solid appearance.
  • Storage: 4°C, protect from light; in solvent: -80°C (6 months), -20°C (1 month).
  • Supplied in milligram-scale quantities suitable for ADC development and analytical work.

Catalog No. 50-002-58373 Supplier Medchemexpress LLC Supplier No. HY145989S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Aminobenzenesulfonic auristatin E-d8 is a deuterium-labeled drug-linker reagent used in antibody-drug conjugate (ADC) research. It combines auristatin E with an aminobenzenesulfonic linker and is supplied as a solid for preclinical, analytical, and linker-optimization studies. It is intended for research use only.

  • Deuterium-labeled version of auristatin E for tracer and stability studies.
  • High purity (98.0%).
  • Chemical formula: C37H56D8N6O8S.
  • Molecular weight: 761.05.
  • White to off-white solid appearance.
  • Storage: 4°C, protect from light; in solvent: -80°C (6 months), -20°C (1 month).
  • Supplied in milligram-scale quantities suitable for ADC development and analytical work.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.