Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Aminobenzenesulfonic auristatin E (TFA) | 1800462-99-0 | MFCD34676939 | 99.4% | 867.03 | C39H65F3N6O10S | 25 MG
SDP

Catalog No. 5000202219
Encompass_Preferred

Aminobenzenesulfonic auristatin E (TFA) is an agent-linker conjugate used in antibody-drug conjugate (ADC) research with potent antitumor activity. It is supplied as the trifluoroacetic acid (TFA) salt in a white to off-white solid and is intended for research applications in oncology and ADC development.

  • Agent-linker conjugate for ADC research.
  • Potent antitumor activity derived from auristatin E.
  • TFA salt; white to off-white solid appearance.
  • High purity (99.4%).
  • Molecular weight 867.03 and formula C39H65F3N6O10S.

Catalog No. 50-002-02219 Supplier Medchemexpress LLC Supplier No. HY145989A25MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Aminobenzenesulfonic auristatin E (TFA) is an agent-linker conjugate used in antibody-drug conjugate (ADC) research with potent antitumor activity. It is supplied as the trifluoroacetic acid (TFA) salt in a white to off-white solid and is intended for research applications in oncology and ADC development.

  • Agent-linker conjugate for ADC research.
  • Potent antitumor activity derived from auristatin E.
  • TFA salt; white to off-white solid appearance.
  • High purity (99.4%).
  • Molecular weight 867.03 and formula C39H65F3N6O10S.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.