Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Aminobenzenesulfonic auristatin E TFA | 867.03 | C39H65F3N6O10S | 5 MG
SDP

Catalog No. 5000202054
Encompass_Preferred

Aminobenzenesulfonic auristatin E TFA is an antibody-drug conjugate (ADC) linker-payload reagent composed of auristatin E linked via an aminobenzenesulfonic ADC linker and supplied as the trifluoroacetate (TFA) salt for preclinical research and ADC development.

  • Agent-linker conjugate for ADC development
  • Contains auristatin E payload, a cytotoxic tubulin modifier
  • Supplied as the trifluoroacetate (TFA) salt
  • Molecular weight 867.03; formula C39H65F3N6O10S
  • Available in research pack sizes including 5 mg

Catalog No. 50-002-02054 Supplier Medchemexpress LLC Supplier No. HY145989A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Aminobenzenesulfonic auristatin E TFA is an antibody-drug conjugate (ADC) linker-payload reagent composed of auristatin E linked via an aminobenzenesulfonic ADC linker and supplied as the trifluoroacetate (TFA) salt for preclinical research and ADC development.

  • Agent-linker conjugate for ADC development
  • Contains auristatin E payload, a cytotoxic tubulin modifier
  • Supplied as the trifluoroacetate (TFA) salt
  • Molecular weight 867.03; formula C39H65F3N6O10S
  • Available in research pack sizes including 5 mg

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.