Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Aminobenzenesulfonic auristatin E TFA | 99.4% | 867.03 | C39H65F3N6O10S | 50 MG
SDP

Catalog No. 5000202288
Encompass_Preferred

Aminobenzenesulfonic auristatin E (TFA) is a drug-linker conjugate intended for antibody-drug conjugate (ADC) research. Supplied as the trifluoroacetic acid (TFA) salt, it targets microtubule/tubulin and is provided in multiple package sizes for research applications.

  • Drug-linker conjugate for ADC-related research.
  • Targets microtubule/tubulin.
  • Reported purity 99.39%.
  • Molecular weight 867.03; formula C39H65F3N6O10S.
  • Available in multiple sizes from 5 mg to 500 mg.

Catalog No. 50-002-02288 Supplier Medchemexpress LLC Supplier No. HY145989A50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Aminobenzenesulfonic auristatin E (TFA) is a drug-linker conjugate intended for antibody-drug conjugate (ADC) research. Supplied as the trifluoroacetic acid (TFA) salt, it targets microtubule/tubulin and is provided in multiple package sizes for research applications.

  • Drug-linker conjugate for ADC-related research.
  • Targets microtubule/tubulin.
  • Reported purity 99.39%.
  • Molecular weight 867.03; formula C39H65F3N6O10S.
  • Available in multiple sizes from 5 mg to 500 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.