Learn More
Eurofins Discoverx ɣ-Aminobutyric acid, 10 mg. (92-1218)
Supplier:  Eurofins Discoverx 921218
Encompass
ɣ-Aminobutyric acid (GABA) is the chief inhibitory neurotransmitter of the CNS. Reduces neuronal excitability, regulates muscle tone, relieves anxiety, promotes lean muscle growth, burns fat, stabilizes blood pressure, and improves mood. Used to reduce symptoms of premenstrual syndrome (PMS), and in treating attention deficit hyperactivity disorder (ADHD). Agonist of the γ-aminobutyric acid receptors GABAA and GABAB. Provided as 10 mg of dry powder. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9742