Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

γ-Aminobutyric acid, ≥97.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014621 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4621 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4621 Supplier MilliporeSigma Supelco Supplier No. 03835250MG
Add to Cart
Add to Cart

γ-Aminobutyric acid is a chief inhibitory neurotransmitter, which is found in the cerebellum, hypothalamus, thalamus and hippocampus. It is formed via the decarboxylation of L-glutamate catalyzed by the enzyme, glutamic acid decarboxylase(GAD).

Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 56-12-2
Linear Formula NH2(CH2)3 COOH
MDL Number MFCD00008226
Melting Point 195°C
Synonym 3-Carboxypropylamine; 4-Aminobutanoic acid; GABA; Piperidic acid; Piperidinic acid
Type Analytical Standard
Quantity 250 mg
Formula Weight 103.12
Percent Purity ≥97.0% (HPLC)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.