Learn More
Revvity Health Sciences Inc Aminobutyric Acid (GABA) γ-[2,3-3H(N)]-, Specific Activity: 25-40Ci (925GBq-1.48TBq)/mmol, 250µCi (9.25MBq)
Catalog No. 502814283
Encompass
Aminobutyric Acid (GABA) γ-[2,3-3H(N)]-, Specific Activity: 25-40Ci (925GBq-1.48TBq)/mmol, 250µCi (9.25MBq)
Catalog No. 50-281-4283 Supplier Revvity Health Sciences Inc Supplier No. NET191250UC
Aminobutyric Acid (GABA) γ-[2,3-3H(N)]-, Specific Activity: 25-40Ci (925GBq-1.48TBq)/mmol, 250µCi (9.25MBq)