Learn More
Revvity Health Sciences Inc Aminobutyric Acid (GABA), γ-[2,3-3H(N)]-, Specific Activity: 70-100Ci (2.59-3.70TBq)/mmol, 250µCi (9.25MBq)
Catalog No. 509050994
Encompass
Aminobutyric Acid (GABA), γ-[2,3-3H(N)]-, Specific Activity: 70-100Ci (2.59-3.70TBq)/mmol, 250µCi (9.25MBq)
Catalog No. 50-905-0994 Supplier Revvity Health Sciences Inc Supplier No. NET191X250UC
Aminobutyric Acid (GABA), γ-[2,3-3H(N)]-, Specific Activity: 70-100Ci (2.59-3.70TBq)/mmol, 250µCi (9.25MBq)