Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aminocaproic acid Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111014854 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4854 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4854 Supplier MilliporeSigma Supelco Supplier No. PHR12241G
Add to Cart
Add to Cart
Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

CAS 60-32-2
Physical Form Neat
Linear Formula H2 N(CH2)5 CO2 H
MDL Number MFCD00008238
Melting Point 207°C to 209°C
Synonym 6-Aminocaproic acid; epsilon-Aminocaproic acid; 6-Aminohexanoic acid; EACA
Type Certified Reference Material
Quantity 1 g
Formula Weight 131.17
Grade Certified Reference Material
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.