Learn More
Abcam 5-(2-Aminoethylamino)- 1-naphthalenesulfonic acid, sodium salt (1,5-EDANS, Na+salt), 250MG
Catalog No. 50996069
Encompass_Preferred
MW 288.3 Da, Purity >95%. Water soluble, blue-green fluorogenic labelling reagent for protein and carbohydrate modifications. EDANS-DABCYL fluorophore-quencher pair is commonly used as a resonance energy transfer pair for peptidase substrate (FRET) analysis.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-996-069 Supplier Abcam Supplier No. AB275074250MG
MW 288.3 Da, Purity >95%. Water soluble, blue-green fluorogenic labelling reagent for protein and carbohydrate modifications. EDANS-DABCYL fluorophore-quencher pair is commonly used as a resonance energy transfer pair for peptidase substrate (FRET) analysis.
The product is subject to the following: Abcam Restricted Use Statement