Learn More
Medchemexpress LLC Aminoethyl-SS-propionic acid | 15579-00-7 | 98.0% | 181.28 g/mol | C5H11NO2S2 | 100 MG
SDP
Catalog No. 5000202695
Encompass_Preferred
Aminoethyl-SS-propionic acid is a cleavable disulfide-containing linker reagent used to prepare antibody-drug conjugates (ADCs). It is supplied as a solid powder for research use and enables attachment of payloads to antibodies via a reducible S-S bond.
  • Cleavable disulfide linker for ADC synthesis and conjugation chemistries.
  • High purity solid reagent, suitable for research applications.
  • Provided in multiple package sizes; common quantity: 100 mg.
  • Stable as powder at -20°C for long-term storage; store solutions at -80°C for extended stability.
Catalog No. 50-002-02695 Supplier Medchemexpress LLC Supplier No. HY140096100MG
Aminoethyl-SS-propionic acid is a cleavable disulfide-containing linker reagent used to prepare antibody-drug conjugates (ADCs). It is supplied as a solid powder for research use and enables attachment of payloads to antibodies via a reducible S-S bond.
  • Cleavable disulfide linker for ADC synthesis and conjugation chemistries.
  • High purity solid reagent, suitable for research applications.
  • Provided in multiple package sizes; common quantity: 100 mg.
  • Stable as powder at -20°C for long-term storage; store solutions at -80°C for extended stability.