Learn More
Medchemexpress LLC Aminoethyl-SS-propionic acid | 15579-00-7 | MFCD07357249 | 181.28 g/mol | C5H11NO2S2 | 50 MG
SDP
Catalog No. 5000204047
Encompass_Preferred
Aminoethyl-SS-propionic acid is a cleavable disulfide-containing linker used in the synthesis of antibody-drug conjugates (ADCs). It contains a terminal amine and a carboxylic acid for conjugation, and a reducible disulfide bond that enables intracellular payload release. Intended for research use only.
  • Provides a reducible disulfide linker for intracellular release.
  • Contains amine and carboxylic acid functional groups for conjugation chemistry.
  • Solid compound; molecular formula C5H11NO2S2, molecular weight 181.28 g/mol.
  • Available in multiple package sizes, including 50 MG.
  • For research use only; not for human or clinical applications.
Catalog No. 50-002-04047 Supplier Medchemexpress LLC Supplier No. HY14009650MG
Aminoethyl-SS-propionic acid is a cleavable disulfide-containing linker used in the synthesis of antibody-drug conjugates (ADCs). It contains a terminal amine and a carboxylic acid for conjugation, and a reducible disulfide bond that enables intracellular payload release. Intended for research use only.
  • Provides a reducible disulfide linker for intracellular release.
  • Contains amine and carboxylic acid functional groups for conjugation chemistry.
  • Solid compound; molecular formula C5H11NO2S2, molecular weight 181.28 g/mol.
  • Available in multiple package sizes, including 50 MG.
  • For research use only; not for human or clinical applications.