Learn More
Medchemexpress LLC Aminoethyl-ss-propionic acid | 15579-00-7 | MFCD07357249 | 98.0% | 181.28 g/mol | C5H11NO2S2 | 10 MG
SDP
Catalog No. 5000202649
Encompass_Preferred
Aminoethyl-ss-propionic acid is a cleavable disulfide-containing ADC linker used to conjugate payloads to antibodies and enable intracellular release under reducing conditions. It is supplied as a purified solid for research use and is commonly provided in small-scale quantities for linker synthesis and optimization.
  • Cleavable disulfide linker enabling intracellular payload release.
  • Contains terminal amine and carboxylic acid functional groups.
  • High purity (approximately 98.0%) suitable for research use.
  • Available in mg and g scales, including 10 MG packages.
  • Stable as a powder at -20°C for long-term storage.
Catalog No. 50-002-02649 Supplier Medchemexpress LLC Supplier No. HY14009610MG
Aminoethyl-ss-propionic acid is a cleavable disulfide-containing ADC linker used to conjugate payloads to antibodies and enable intracellular release under reducing conditions. It is supplied as a purified solid for research use and is commonly provided in small-scale quantities for linker synthesis and optimization.
  • Cleavable disulfide linker enabling intracellular payload release.
  • Contains terminal amine and carboxylic acid functional groups.
  • High purity (approximately 98.0%) suitable for research use.
  • Available in mg and g scales, including 10 MG packages.
  • Stable as a powder at -20°C for long-term storage.