Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Aminoethyl-SS-propionic acid | 15579-00-7 | MFCD07357249 | 98.0% | 181.28 g/mol | C5H11NO2S2 | 500 MG
SDP

Catalog No. 5000204164
Encompass_Preferred

Aminoethyl-SS-propionic acid is a cleavable disulfide-containing linker used in the research synthesis of antibody-drug conjugates (ADCs). Supplied as a white to off-white solid with high purity, it enables intracellular release of conjugated payloads upon reduction and is provided in multiple laboratory-scale packaging sizes.

  • Cleavable disulfide-containing linker for ADC synthesis.
  • Formula: C5H11NO2S2.
  • CAS number: 15579-00-7.
  • Molecular weight: 181.28 g/mol.
  • Purity: 98.03% (reported by supplier).
  • Appearance: white to off-white solid.
  • Available in multiple sizes, including 500 mg.

Catalog No. 50-002-04164 Supplier Medchemexpress LLC Supplier No. HY140096500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Aminoethyl-SS-propionic acid is a cleavable disulfide-containing linker used in the research synthesis of antibody-drug conjugates (ADCs). Supplied as a white to off-white solid with high purity, it enables intracellular release of conjugated payloads upon reduction and is provided in multiple laboratory-scale packaging sizes.

  • Cleavable disulfide-containing linker for ADC synthesis.
  • Formula: C5H11NO2S2.
  • CAS number: 15579-00-7.
  • Molecular weight: 181.28 g/mol.
  • Purity: 98.03% (reported by supplier).
  • Appearance: white to off-white solid.
  • Available in multiple sizes, including 500 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.