Learn More
Cayman Chemical AmInoethylsPropIonIc ACD 100mg
SDP
Catalog No. 502910103
Encompass_Preferred
A cleavable disulfide linker; has been used in the synthesis of ADCs
Catalog No. 50-291-0103 Supplier Cayman Chemical Supplier No. 35501100 MG
A cleavable disulfide linker; has been used in the synthesis of ADCs